Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Linear Spring Scales

2000 g x 40 g (20 N x 0.4 N)
Catalog # 470148-882
Supplier:   VWR International
VWR® Linear Spring Scales
Catalog # 470148-882
Supplier:  VWR International
VWR® Linear Spring Scales
VWR® Linear Spring Scales

2000 g x 40 g (20 N x 0.4 N)

Liquidación
$3.35
Oferta

Válido hasta 31/12/2026

Log in Para disponibilidad
Each (1UN) Case de 100 (1UN)
Product Details & Documents

Designed for Heavy Duty Student Use

  • Feature easy to read markings
  • Durable for years of regular student use
  • Length: 14 cm
  • Each scale is easily zeroed

Each has a plated steel suspension ring at the top, and a hook at the bottom.

Especificaciones
  • Weighing Capacity:
    2000 g x 40 g (20 N x 0.4 N)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.