Orders LinkContactUsLinkComponent
Burner Tripods
Tripod, Burner, 23 cm x 13 cm x 9 cm, 5" (3.5" I.D.)
  470148-880
Burner Tripods
  470148-880
Burner Tripods
Burner Tripods
Tripod, Burner, 23 cm x 13 cm x 9 cm, 5" (3.5" I.D.)
$7.20
  31/12/2026
Each (1UN) Case de 20 (1UN)
Good quality, enameled cast iron tripods for supporting small objects over a burner.
  • Cast iron, enameled finish
  • Removable zinc plated legs
Good quality, enameled cast iron tripods for supporting small objects over a burner. The legs detach for easy storage.
  • Ext.Ø:
    5" (3.5" I.D.)
  • Description:
    Tripod, Burner, 23 cm x 13 cm x 9 cm