Burner Tripods
Tripod, Burner, 23 cm x 13 cm x 9 cm, 5" (3.5" I.D.)
470148-880
Good quality, enameled cast iron tripods for supporting small objects over a burner.
- Cast iron, enameled finish
- Removable zinc plated legs
Good quality, enameled cast iron tripods for supporting small objects over a burner. The legs detach for easy storage.
- Ext.Ø:5" (3.5" I.D.)
- Description:Tripod, Burner, 23 cm x 13 cm x 9 cm