Order Entry
Mexico
Orders LinkContactUsLinkComponent
Porous Cups

Porous Cups

Porous cups, 40×90 mm
Catalog # 470148-830
Número de proveedor #:  175933
Porous Cups
Catalog # 470148-830
Porous Cups
Porous Cups

Porous cups, 40×90 mm

Price
$78.48
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents

Porous cylindrical cups.

  • Made of ceramic

Pore diameter is approximately 0.5 µm.

Especificaciones
  • Description:
    Porous cups
  • Diameter ×H:
    40×90 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.