Disposable Micropipet Tips
1 - 200 µl, Yellow, 1000 Tips/Bag
470148-816
High quality micropipette tips to meet any lab's needs.
- Certified DNase, RNase and pyrogen free
- Material: PP
- Autoclavable
- Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
- Volume:1 - 200 µl
- Tip Color:Yellow
- Packed:1000 Tips/Bag