Lil'pet Micropipette
5 µl
Catalog # 470148-804
Supplier: VWR International
Jump to:
Product Details & Documents
Lil'pet Micropipettes feature ideal volumes for gel loading.
- Units accept standard micropipette tips
- Economical
- Durable
- Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
Especificaciones
- Volume:5 µl