Orders LinkContactUsLinkComponent
VWR® Volumetric Flasks
1000 ml
  470148-798
VWR® Volumetric Flasks
  470148-798
VWR® Volumetric Flasks
VWR® Volumetric Flasks
1000 ml
$390.74
Each (1UN) Case de 6 (1UN)
Our borosilicate flasks meet ASTM standards.
  • Class A Flasks
  • Borosilicate Glass Construction
  • Graduated Marking for Measurements
  • Economical
  • Snap-Caps Included
The necks fit the included snap-caps.
The Class A, volumetric flasks include a graduation line for accurate measurements.
  • Capacity:
    1000 ml