Orders LinkContactUsLinkComponent
Adjustable Bunsen Burners
Adjustable Bunsen Burners
Gas burner, LPG, 2000 - 3200 BTU/cu. ft.
Catalog # 470148-690
Adjustable Bunsen Burners
Catalog # 470148-690
Supplier:  
Adjustable Bunsen Burners
Adjustable Bunsen Burners
Gas burner, LPG, 2000 - 3200 BTU/cu. ft.
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
VWR BURNER BUNSEN ART GAS Catalog #89038-528
El producto 89038-528 es un reemplazo para 470148-690 y tiene el mismo embalaje y especificaciones, pero puede tener una cantidad diferente.
Product Details & Documents
Precise Flame Control.
  • Gas Inlet: 5/16" O.D.
  • Mixing Tube: 1/2" O.D.
  • Height: 63/8"
These are similar to the standard burner, but with a fine pitch needle valve for flame adjustment and aluminum mixing tube with flame stabilizer.
Documents
Especificaciones
  • Description:
    Gas burner
  • For:
    LPG
  • BTU Output:
    2000 - 3200 BTU/cu. ft.