Order Entry
Mexico
Orders LinkContactUsLinkComponent
Black Rubber Bib Apron

Black Rubber Bib Apron

Black, S, 30×24"
Catalog # 470148-674
Supplier #:  RBAP-CH
Black Rubber Bib Apron
Catalog # 470148-674
Black Rubber Bib Apron
Black Rubber Bib Apron

Black, S, 30×24"

Price
$109.11
Log in for availability
Each (1UN) Case de 25 (1UN)
Product Details & Documents

Supply Your Lab with Personal Protective Equipment

  • Basic black rubber apron
  • Perfect for labs, kitchens, or shops
  • Contains latex

Suitable for general lab work. Comfortable and flexible Black Rubber Bib Aprons are waterproof and resistant to most chemicals used by students in the lab.

Specifications
  • Clothing Size:
    S
  • Color:
    Black
  • L×W:
    30×24"
  • Cat. No.:
    470148-674
  • Garment application:
    Food|Laboratory
  • Gender:
    Unisex
  • Fabric:
    Rubber
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.