Vinyl Lab Apron
Child Apron, Clear, 27×36"
Catalog # 470148-664
Supplier: ELECTRONIC INSTRUMENTATION
Supplier #: VNAP-CH
Jump to:
Product Details & Documents
Tough vinyl lab aprons are resistant to lab reagents, acids, alkalis, oils, and grease.
- Halter Design
- Aprons Do Not Restrict Movement
- Two Sizes Available
- Resistant to Acids, Alkalis, Oils, and More
- Reinforced Strain Points
- Latex free
The halter design keeps arms free for unrestricted movement in the lab.
The aprons are reinforced at strain points.
Especificaciones
- Color:Clear
- Description:Child Apron
- W×L:27×36"