Orders LinkContactUsLinkComponent
Vinyl Lab Apron
Vinyl Lab Apron
Child Apron, Clear, 27×36"
Catalog # 470148-664
Supplier #:  VNAP-CH
Vinyl Lab Apron
Catalog # 470148-664
Vinyl Lab Apron
Vinyl Lab Apron
Child Apron, Clear, 27×36"
Price
$48.37
Log in Para disponibilidad
Each (1UN) Case de 50 (1UN)
Product Details & Documents
Tough vinyl lab aprons are resistant to lab reagents, acids, alkalis, oils, and grease.
  • Halter Design
  • Aprons Do Not Restrict Movement
  • Two Sizes Available
  • Resistant to Acids, Alkalis, Oils, and More
  • Reinforced Strain Points
  • Latex free
The halter design keeps arms free for unrestricted movement in the lab.
The aprons are reinforced at strain points.
Especificaciones
  • Color:
    Clear
  • Description:
    Child Apron
  • W×L:
    27×36"