Tirrill Burners
Tirrill burner, 800 - 1200 BTU/cu. ft.
Catalog # 470148-642
Supplier: VWR International
Jump to:
Product Details & Documents
Precise Flame Control.
- Gas Inlet: 3/8" O.D.
- Mixing Tube: 1/2" O.D.
- Height: 51/4"
- LP gas burner, compatible with propane
These burners allow you to adjust both the gas and air supply for precise flame control. The aluminum mixing tube features a flame stabilizer, as well as both needle valve and air ports. It has a nickel-plated zinc base.
Especificaciones
- Description:Tirrill burner
- BTU Output:800 - 1200 BTU/cu. ft.