Orders LinkContactUsLinkComponent
Tirrill Burners
Tirrill Burners
Tirrill burner, 800 - 1200 BTU/cu. ft.
Catalog # 470148-642
Supplier:   VWR International
Tirrill Burners
Catalog # 470148-642
Supplier:  VWR International
Tirrill Burners
Tirrill Burners
Tirrill burner, 800 - 1200 BTU/cu. ft.
Price
$807.43
Log in Para disponibilidad
Each (1UN) Case de 10 (1UN)
Product Details & Documents
Precise Flame Control.
  • Gas Inlet: 3/8" O.D.
  • Mixing Tube: 1/2" O.D.
  • Height: 51/4"
  • LP gas burner, compatible with propane
These burners allow you to adjust both the gas and air supply for precise flame control. The aluminum mixing tube features a flame stabilizer, as well as both needle valve and air ports. It has a nickel-plated zinc base.
Especificaciones
  • Description:
    Tirrill burner
  • BTU Output:
    800 - 1200 BTU/cu. ft.