Polypropylene Jars
Polypropylene Jars, 250 ml
Catalog # 470147-472
Supplier: DYNALON
Número de proveedor #: 421145
Jump to:
Product Details & Documents
Convenient for Storing and Collecting Lab Supplies and Specimens.
- Supplied with linerless polypropylene screw caps
- Leak-proof
- Autoclavable
Especificaciones
- Capacity:250 ml
- Description:Polypropylene Jars