Orders LinkContactUsLinkComponent
VWR® Micropipette Tip Reload System
VWR® Micropipette Tip Reload System
100 - 1250 µl, Natural, 1000 Tips/Bag, 5 Bags/Case
Catalog # 470146-156
Número de proveedor #:  112NXL
VWR® Micropipette Tip Reload System
Catalog # 470146-156
VWR® Micropipette Tip Reload System
VWR® Micropipette Tip Reload System
100 - 1250 µl, Natural, 1000 Tips/Bag, 5 Bags/Case
Price
$921.00
Log in Para disponibilidad
Paquete de 1,000 Case de 10 (1,000,UN)
Product Details & Documents
Save time, plastic, and money with the Micropipette tip reload system.
  • Tips available in three different configuarations
  • Save plastic and re-use VWR® hinged rack boxes with the Reload Sleeves
  • Flip-lock boxes offer easy one-handed operation
  • Lid lip enables stable stacking
  • Tips work with most manufacturer's micropipettors
The reloading operation features sleeve alignment, which orients loading for a perfect fit every time.
The tongue and groove format keeps the insert fastened while applying tips, and the tall clip makes the insert easy to handle without touching tips with your hand.
Packaging: The Racked Tips contain 10 boxes ('racks') of 96 per pack. There are 5 packs per case.
The Reload Sleeves contain 10 inserts of 96 tips per pack. There are 5 packs per case.
The Bulk Bags contain 1000 loose tips per bag. There are 10 bags per case.
Documents
Especificaciones
  • Volume:
    100 - 1250 µl
  • Tip Color:
    Natural
  • Packed:
    1000 Tips/Bag, 5 Bags/Case