Compartmented Plastic Boxes, Clear, Flambeau
18, 15.7 cm (6³/₁₆")×26.7 cm (10¹/₂")×4 cm (1⁹/₁₆") (W×L×H)
470145-406
: K618
Keep small specimens or other items organized and visible in these clear polystyrene boxes with hinged lids.
- Transparent
- Butadiene copolymer material
- Dimensions:15.7 cm (6³/₁₆")×26.7 cm (10¹/₂")×4 cm (1⁹/₁₆") (W×L×H)
- No. of Compartments:18