Orders LinkContactUsLinkComponent
Pipette Pumps
Pipette Pumps
1 & 2 ml Pipette Pump (Blue)
Catalog # 470145-298
Supplier:   VWR International
Pipette Pumps
Catalog # 470145-298
Supplier:  VWR International
Pipette Pumps
Pipette Pumps
1 & 2 ml Pipette Pump (Blue)
Price
$834.00
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents
Safe and accurate.
  • Easily disassembled for cleaning
  • Color-coded for pipette size selection
  • Resist acid and alkalis
These pumps are easy to use in one-handed operation. The thumbwheel is simply rotated for precise filling or dispensing. Measuring pipettes fit smoothly into tapered chuck and a plunger may be used for quick dispensation.
Especificaciones
  • Description:
    1 & 2 ml Pipette Pump (Blue)