Pipette Pumps
1 & 2 ml Pipette Pump (Blue)
Catalog # 470145-298
Supplier: VWR International
Jump to:
Product Details & Documents
Safe and accurate.
- Easily disassembled for cleaning
- Color-coded for pipette size selection
- Resist acid and alkalis
These pumps are easy to use in one-handed operation. The thumbwheel is simply rotated for precise filling or dispensing. Measuring pipettes fit smoothly into tapered chuck and a plunger may be used for quick dispensation.
Especificaciones
- Description:1 & 2 ml Pipette Pump (Blue)