Orders LinkContactUsLinkComponent
Pipette Pumps
Pipette Pumps
5 & 10 ml Pipette Pump (Green)
Catalog # 470145-296
Supplier:   VWR International
Pipette Pumps
Catalog # 470145-296
Supplier:  VWR International
Pipette Pumps
Pipette Pumps
5 & 10 ml Pipette Pump (Green)
Price
$894.00
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents
Safe and accurate.
  • Easily disassembled for cleaning
  • Color-coded for pipette size selection
  • Resist acid and alkalis
These pumps are easy to use in one-handed operation. The thumbwheel is simply rotated for precise filling or dispensing. Measuring pipettes fit smoothly into tapered chuck and a plunger may be used for quick dispensation.
Especificaciones
  • Description:
    5 & 10 ml Pipette Pump (Green)