Orders LinkContactUsLinkComponent
Pipette Pumps
25 ml Pipette Pump (Red)
Catalog # 470145-294
Supplier:   VWR International
Pipette Pumps
Catalog # 470145-294
Supplier:  VWR International
Pipette Pumps
Pipette Pumps
25 ml Pipette Pump (Red)
Price
$1,140.00
Log in for availability
Each (1UN) Case de 12 (1UN)
Product Details & Documents
Safe and accurate.
  • Easily disassembled for cleaning
  • Color-coded for pipette size selection
  • Resist acid and alkalis
These pumps are easy to use in one-handed operation. The thumbwheel is simply rotated for precise filling or dispensing. Measuring pipettes fit smoothly into tapered chuck and a plunger may be used for quick dispensation.
Specifications
  • Description:
    25 ml Pipette Pump (Red)