Compartmented Plastic Boxes, Clear, Flambeau
24, 21.6 cm (8¹/₂")×32.4 cm (12³/₄")×5.4 cm (2¹/₈") (W×L×H)
470145-018
: A824
Keep small specimens or other items organized and visible in these clear polystyrene boxes with hinged lids.
- Transparent
- Butadiene copolymer material
- Dimensions:21.6 cm (8¹/₂")×32.4 cm (12³/₄")×5.4 cm (2¹/₈") (W×L×H)
- No. of Compartments:24