Orders LinkContactUsLinkComponent
Specific Gravity Set
Specific Gravity Set
Specific Gravity Set, Copper Cylinder Only
Catalog # 470137-634
Supplier:   VWR International
Specific Gravity Set
Catalog # 470137-634
Supplier:  VWR International
Specific Gravity Set
Specific Gravity Set
Specific Gravity Set, Copper Cylinder Only
Liquidación
$4.05
Oferta
Válido hasta 31/12/2026
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Teach About Gravity
  • Metals included are: aluminum, brass, copper and steel
  • Each cylinder is 5cm long and 13mm diameter
  • Purchase as a set or individually
Determine the gravity of common metals using this set of specific gravity cylinders. Perfect for specific gravity determinations in the classroom. 
Caution: WARNING: CHOKING HAZARD-Not for children under 3 years.
Especificaciones
  • Description:
    Specific Gravity Set, Copper Cylinder Only