Specific Gravity Set
Specific Gravity Set, Aluminum Cylinder Only
Catalog # 470137-630
Supplier: VWR International
Jump to:
Product Details & Documents
Teach About Gravity
- Metals included are: aluminum, brass, copper and steel
- Each cylinder is 5cm long and 13mm diameter
- Purchase as a set or individually
Determine the gravity of common metals using this set of specific gravity cylinders. Perfect for specific gravity determinations in the classroom.
Caution: WARNING: CHOKING HAZARD-Not for children under 3 years.
Especificaciones
- Description:Specific Gravity Set, Aluminum Cylinder Only
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



