Specific Gravity Set
Specific Gravity Set, Set of 4
470137-628
Teach About Gravity
- Metals included are: aluminum, brass, copper and steel
- Each cylinder is 5cm long and 13mm diameter
- Purchase as a set or individually
Determine the gravity of common metals using this set of specific gravity cylinders. Perfect for specific gravity determinations in the classroom.
: WARNING: CHOKING HAZARD-Not for children under 3 years.
- Description:Specific Gravity Set, Set of 4