Orders LinkContactUsLinkComponent
Bottle Top Dispenser
Bottle Top Dispenser
Bottle top dispenser, 2.5-30 ml
Catalog # 470134-948
Supplier:   VWR International
Bottle Top Dispenser
Catalog # 470134-948
Supplier:  VWR International
Bottle Top Dispenser
Bottle Top Dispenser
Bottle top dispenser, 2.5-30 ml
Price
$5,812.99
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Spill-Free and Safe Dispensing
Prevent spills while making your laboratory a cleaner and safer environment. This innovative bottle top dispensing system allows for use with a wide range of laboratory chemicals. The unique nozzle easily dispenses liquids accurately into the receiving container and is autoclavable to 121˚ C. The system includes a variety of bottle adapters to fit standard-size laboratory bottles. Graduated reagent bottles are recommended for replacements.
Especificaciones
  • Description:
    Bottle top dispenser, 2.5-30 ml