Science Take-Out® Beadle Plasticus Evolution
Beadle Plasticus evolution single kit, 1
470127-842
: STO-125
How Do Different Environments Affect A Population?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
How do different environments affect a population? Investigate the changes in a population of "Beadle plasticus" in different environments. Model how the environment affects the frequencies of "Beadle" characteristics. This complete "dry lab" activity contains all required materials.
- Description:Beadle Plasticus evolution single kit
- No. of Lab Groups:1