Watchmaker Forceps
Economy Watchmaker Forceps
Catalog # 470126-100
Supplier: DUNRITE INSTRUMENTS SE
Número de proveedor #: 141400
Jump to:
Product Details & Documents
Ultra fine control.
- Super fine point for precise work
- Smooth surface prevents damage to delicate tissues
- Broad handle for easy control
This is the traditional instrument for fine dissection work, with fine points and a broad handle. Choose between the standard broad handled, and economy narrow handled versions, both with very sharp, smooth tips.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Economy Watchmaker Forceps