Orders LinkContactUsLinkComponent
Glass Plates
Glass Plates
Glass Plates, Type: Clear, Size: 6" x 6"
Catalog # 470123-230
Supplier:   VWR International
Glass Plates
Catalog # 470123-230
Supplier:  VWR International
Glass Plates
Glass Plates
Glass Plates, Type: Clear, Size: 6" x 6"
Price
$473.32
Log in Para disponibilidad
Paquete de 12
Product Details & Documents
Plates made of soda lime glass, 2mm thick, with ground polished edges. Useful for a variety of general laboratory applications including as a cover, mixing surface, etc.
  • Soda lime glass
  • Polished edges
Clean cut edges, not ground. Available in Clear or Cobalt Blue.
Especificaciones
  • Description:
    Glass Plates, Type: Clear, Size: 6" x 6"