Glass Plates
Glass Plates, Type: Clear, Size: 4" x 4"
470123-228
Plates made of soda lime glass, 2mm thick, with ground polished edges. Useful for a variety of general laboratory applications including as a cover, mixing surface, etc.
- Soda lime glass
- Polished edges
Clean cut edges, not ground. Available in Clear or Cobalt Blue.
- Description:Glass Plates, Type: Clear, Size: 4" x 4"