Orders LinkContactUsLinkComponent
Glass Plates
Glass Plates, Type: Clear, Size: 3" x 3"
  470123-226
Glass Plates
  470123-226
Glass Plates
Glass Plates
Glass Plates, Type: Clear, Size: 3" x 3"
$300.60
Paquete de 12
Plates made of soda lime glass, 2mm thick, with ground polished edges. Useful for a variety of general laboratory applications including as a cover, mixing surface, etc.
  • Soda lime glass
  • Polished edges
Clean cut edges, not ground. Available in Clear or Cobalt Blue.
  • Description:
    Glass Plates, Type: Clear, Size: 3" x 3"