Orders LinkContactUsLinkComponent
Glass Plates
Glass Plates
Glass Plates, Type: Clear, Size: 2" x 2"
Catalog # 470123-224
Número de proveedor #:  GLP2X2-PK/12
Glass Plates
Catalog # 470123-224
Supplier:  
Glass Plates
Glass Plates
Glass Plates, Type: Clear, Size: 2" x 2"
Discontinued
Product Details & Documents
Plates made of soda lime glass, 2mm thick, with ground polished edges. Useful for a variety of general laboratory applications including as a cover, mixing surface, etc.
  • Soda lime glass
  • Polished edges
Clean cut edges, not ground. Available in Clear or Cobalt Blue.
Especificaciones
  • Description:
    Glass Plates, Type: Clear, Size: 2" x 2"