Levitating Magnets Set
Levitating Magnets Set, Replacement Magnets
Catalog # 470122-660
Supplier: DOWLING MAGNETS,LLC
Número de proveedor #: CS-100
Jump to:
Product Details & Documents
Floating Magnets Seem to Defy Gravity
- Includes five brightly colored ceramic ring magnets each 28mm in diameter and a 15cm wooden dowel on which to mount them
Bring a little levitation to your classroom with these ring magnets. They graphically demonstrate both attraction and repulsion of magnetism. Replacement magnet available.
Especificaciones
- Description:Levitating Magnets Set, Replacement Magnets