Order Entry
Mexico
Orders LinkContactUsLinkComponent
Lens Paper

Lens Paper

Lens Paper, Form: Loose Sheets, Sheets, 12", 9"
Catalog # 470122-632
Supplier:   RAYLABCON INC SE
Supplier #:  20 20 5103
Lens Paper
Catalog # 470122-632
Supplier:  RAYLABCON INC SE
Lens Paper
Lens Paper

Lens Paper, Form: Loose Sheets, Sheets, 12", 9"

Price
$140.20
Log in for availability
Paquete de 50
Product Details & Documents

This soft, absorbent paper cleans lenses and optical elements.

  • Available in booklet and sheet form

The paper removes grit, lint, and dirt from cleaning lenses and optical elements.

Specifications
  • Width:
    9"
  • Description:
    Lens Paper, Form: Loose Sheets, Sheets
  • Length:
    12"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.