Orders LinkContactUsLinkComponent
Label Tape, White
Label Tape, White
White Label Tape, Width: 1"
Catalog # 470122-606
Número de proveedor #:  ST-10-1
Label Tape, White
Catalog # 470122-606
Label Tape, White
Label Tape, White
White Label Tape, Width: 1"
Price
$442.93
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
General purpose tape which adheres to nearly any surface.
  • Aligned with GLOBE protocols
  • Length: 41¹/₂'
Water and oil proof. Can be marked with pen, pencil, and more.
Especificaciones
  • Description:
    White Label Tape, Width: 1"