Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Stackable Banana Plug Connecting Wires

Black Stackable Banana Plug Connecting Wires, 36"
  470122-136
Stackable Banana Plug Connecting Wires
  470122-136
Stackable Banana Plug Connecting Wires
Stackable Banana Plug Connecting Wires

Black Stackable Banana Plug Connecting Wires, 36"

 
$58.62
   
Each (1UN)
 

Flexible, Insulated Wire.

  • Insulated 4 mm banana plug at both ends
  • Plated spring contacts
  • Stack to accept other banana plugs
  • With side connectors
 
  • Description:
    Black Stackable Banana Plug Connecting Wires
  • Length:
    36"