Economy Dissection Instruments
Student Scalpel, Nickel Plated, 128 mm (5")
Catalog # 470122-000
Supplier: DUNRITE INSTRUMENTS SE
Número de proveedor #: WLS26640
Jump to:
Product Details & Documents
Priced for even the tightest budget.
- Plated steel
- Available in multiple sizes and styles
- Great for student dissection sets
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Student Scalpel
- Length:128 mm (5")
- Material:Nickel Plated