Orders LinkContactUsLinkComponent
XXXX Quality Corks
XXXX Quality Corks
12
Catalog # 470121-848
   Low Manufacturing Impact
Supplier #:  55512
XXXX Quality Corks
Catalog # 470121-848
   Low Manufacturing Impact
XXXX Quality Corks
XXXX Quality Corks
12
Price
$496.23
Log in for availability
Paquete de 50
Product Details & Documents
XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
  • Fine Grain
  • Ideal for Use as Tight-Fitting Stoppers
  • Common Sizes
  • Regular Length
The listed sizes listed are common in school science laboratories.
The corks are regular length.
Caution: Product contains latex. May cause allergic reactions.
Specifications
  • Stopper No.:
    12
  • Cat. No.:
    470121-848
  • Environmentally Preferable:
    Low Manufacturing Impact