Resistance Blocks
100 Ohm Resistance block
Catalog # 470121-098
Supplier: VWR International
Jump to:
Product Details & Documents
Economical yet Sturdy
- Simple to use
- Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
Specifications
- Description:100 Ohm Resistance block