Orders LinkContactUsLinkComponent
VWR® Volumetric Flasks, Class B
VWR® Volumetric Flasks, Class B
VWR® Volumetric Flasks — Class B, Cap., mL: 1000, Tolerance, ±mL: 0.60
Catalog # 470120-672
Supplier:   VWR International
VWR® Volumetric Flasks, Class B
Catalog # 470120-672
Supplier:  VWR International
VWR® Volumetric Flasks, Class B
VWR® Volumetric Flasks — Class B, Cap., mL: 1000, Tolerance, ±mL: 0.60
Price
$2,140.56
Log in Para disponibilidad
Paquete de 6
Product Details & Documents
Our new high-quality borosilicate volumetric flasks meet ASTM standards and are a great value.
  • Borosilicate glass construction
  • Snap-on caps
  • Graduated line for accurate measurements
Class B with capacity tolerances twice those specified for Class A volumetric work. The necks are tooled to fit the included snap-caps. With graduation line for accurate measurements.
Especificaciones
  • Description:
    VWR® Volumetric Flasks — Class B, Cap., mL: 1000, Tolerance, ±mL: 0.60