Orders LinkContactUsLinkComponent
Acrylic Buret
Acrylic Buret
100 ml, 0.2 ml
Catalog # 470111-676
Número de proveedor #:  11503
Acrylic Buret
Catalog # 470111-676
Acrylic Buret
Acrylic Buret
100 ml, 0.2 ml
Price
$617.77
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
These hardy burets are ideal for classroom use.
  • Acrylic
  • Transparent
  • Leakproof Stopcock Included
  • Burets Unaffected by Most Dilute Aqueous Solutions
  • Heavy-Duty
The transparent, acrylic burets are supplied with self-lubricating, leakproof PMP stopcock.
They are unaffected by most dilute aqueous solutions except ammonia and HF and are not recommended for use with ethanol or organic solvents.
Especificaciones
  • Graduation Interval:
    0.2 ml
  • Capacity:
    100 ml