Blocks and Refraction Plates
Blocks and Refraction Plates, Clear Glass Block, 114 x 63 x 19mm
Catalog # 470110-632
Supplier: VWR International
Jump to:
Product Details & Documents
Parallel-Sided Blocks for Ray Optics
Ideal for refraction and internal reflection studies as well as index of refraction measurements, these clear blocks are free of bubbles and streaks and have polished surfaces. Acrylic blocks and refraction plates have frosted ends, glass block is completely clear.
Especificaciones
- Description:Blocks and Refraction Plates, Clear Glass Block, 114 x 63 x 19mm