Marina® Easycatch Nylon Nets
Dip Net, 8", 6"
470106-314
: 11267
Marina easy catch fish nets make managing and handling fish very easy and convenient.
Made with loosely-weaved nylon mesh, the nets allow easy, unhindered movement with minimum water resistance. All Easy Catch Fish Nets have vinyl-coated handles that ensure a safe and comfortable grip.
- Width:6"
- Description:Dip Net
- Length:8"