Vinyl Tape
Vinyl Tape, Blue
Catalog # 470043-238
Supplier: GRAYBAR ELECTRIC CO INC CP
Número de proveedor #: 95011182
Jump to:
Product Details & Documents
General use vinyl tape is availabe in blue or green.
Tape measures ³/₄"W × 66'L.
Especificaciones
- Description:Vinyl Tape, Blue