Orders LinkContactUsLinkComponent
Screw Compression Clamp with Open Jaw
Screw Compression Clamp with Open Jaw, Jaw Opening, mm: 13 to 19
  470032-730
 :  HSCOP1
Screw Compression Clamp with Open Jaw
  470032-730
Screw Compression Clamp with Open Jaw
Screw Compression Clamp with Open Jaw
Screw Compression Clamp with Open Jaw, Jaw Opening, mm: 13 to 19
$47.98
Each (1UN)
Easily clamp tubes up to 1" in diameter.
  • Zinc-plated brass
Can use as a closed clamp or open.
  • Description:
    Screw Compression Clamp with Open Jaw, Jaw Opening, mm: 13 to 19