Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Somso® Gorilla Skull

Gorilla Skull, Female, 2, 15 cm, 27.5 cm, 15.5 cm
  470029-320
 :  ZOS 51
Somso® Gorilla Skull
  470029-320
Somso® Gorilla Skull
Somso® Gorilla Skull

Gorilla Skull, Female, 2, 15 cm, 27.5 cm, 15.5 cm

 
$9,340.00
   
Each (1UN)
 

Compare the structural differences between male and female gorillas (Gorilla gorilla), noting the smaller overall size, and size of central crest and brow ridges, of the female, by using this unmounted plastic, adult female skull with the SOMSO male gorilla skull. Both feature a removable lower jaw.

  • Cast from real specimens
  • Male and female versions available for comparison
  • 5 year manufacturers warranty
 
  • Description:
    Gorilla Skull, Female
  • Height:
    15.5 cm
  • Length:
    27.5 cm
  • Width:
    15 cm
  • No. of Parts:
    2