Orders LinkContactUsLinkComponent
Fluorite (Crystalline)
Fluorite (Crystalline)
Fluorite mineral display, Average 2×3"
Catalog # 470026-390
Fluorite (Crystalline)
Catalog # 470026-390
Supplier:  
Fluorite (Crystalline)
Fluorite (Crystalline)
Fluorite mineral display, Average 2×3"
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLUORITE XL GROUP 2 X2 -4 X4 CUBIC MX Catalog #470226-654
Product 470226-654 is a replacement for 470026-390 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Deep purple, cubic crystal cluster.
  • Purple color
  • Mexico
Specifications
  • Description:
    Fluorite mineral display
  • L×W:
    Average 2×3"