Ward's® Artificial Urine
Artificial urine with glucose, Laboratory, 500 ml
Supplier: ALDON CORP SE
Supplier #: UU0080-500
Jump to:
Product Details & Documents
Completely safe and realistic for urinalysis.
Students may find urology testing demonstrations or lab exercises more pleasant if they do not have to use the real thing.
Caution: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Especificaciones
- Description:Artificial urine with glucose
- Size:500 ml
- Grade:Laboratory