Orders LinkContactUsLinkComponent
Pocket Magnifiers
Pocket Magnifiers
Pocket Magnifier Single Lens
Catalog # 470021-802
Número de proveedor #:  23201-3
Pocket Magnifiers
Catalog # 470021-802
Pocket Magnifiers
Pocket Magnifiers
Pocket Magnifier Single Lens
Price
$52.76
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
These pocket magnifiers are economical enough to outfit an entire class.
  • Handy plastic magnifiers
  • Ideal for younger students
  • Folds conveniently into attached case
  • Double/triple lenses can be stacked to increase magnification
Handy plastic magnifiers, with your choice of single, double, or triple lenses, are ideal for younger students. Each folds conveniently into an attached oval case. The 10X and 15X models’ double and triple lenses can be used separately or “stacked” to increase the magnification.
Dimensions: 1 1/2L x 1 1/4W x 1/4"H folded, 2 3/4"L extended.
Especificaciones
  • Magnification:
  • Description:
    Pocket Magnifier Single Lens