Medium Grade, 15 μm
Filter Paper, Medium Grade, 90 mm diameter
Catalog # 470019-882
Supplier: UNITED SCIENTIFIC SUPPLIES
: FPR009
Jump to:
Product Details & Documents
The most widely used qualitative filter paper for routine applications.
- Medium grade, 15 μm
Holds flocculants and precipitates commonly encountered in laboratory exercises.
: Packages of 100.
Especificaciones
- Description:Filter Paper, Medium Grade, 90 mm diameter