Rectangular Supports
4×6", ⁵/₁₆", 18"
Catalog # 470019-526
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: SSB4X6-CI
Jump to:
Product Details & Documents
Tip-free stability for experiments.
- Glassware can sit directly on base
- Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.
Especificaciones
- Rod Length:18"
- Base Dimensions:4×6"
- Rod Diameter:⁵/₁₆"