Orders LinkContactUsLinkComponent
Cellophane
Cellophane
Cellophane Roll, Red, 20×100"
Catalog # 470019-312
Supplier #:  71002
Cellophane
Catalog # 470019-312
Cellophane
Cellophane
Cellophane Roll, Red, 20×100"
Price
$572.99
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Use the various colors to demonstrate plants’ reaction to light waves.
  • Made from recycled materials
Also useful for demonstrating the effects of mixing various colors of light.
Especificaciones
  • Color:
    Red
  • Description:
    Cellophane Roll
  • Size:
    20×100"