Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Glass Depression Slides

Single concavity slides, 3 mm, 3×1"
Catalog # 470019-022
 :  CSTK01
Glass Depression Slides
Catalog # 470019-022
Glass Depression Slides
Glass Depression Slides

Single concavity slides, 3 mm, 3×1"

Price
$216.95
Log in Para disponibilidad
Paquete de 12
Product Details & Documents

These depression slides are ideal for viewing living specimens suspended in liquid.

  • Made with quality sheet glass with ground edges
  • Sold in boxes of 12 slides
  • Available with single or double concavities

These precleaned slides have a round, shallow depression in the center of the slide, making it easy to view materials in liquid.

Cavities measure approximately 16mm in diameter, and 0.8mm deep.

Especificaciones
  • Description:
    Single concavity slides
  • L×W:
    3×1"
  • Thickness:
    3 mm