Rectangular Glass Coverslips
Glass cover slips, rectangular, 40×22 mm, 0.15 mm, #1
Catalog # 470019-008
Supplier: C & A SCIENTIFIC CO INC
Número de proveedor #: 94-2240
Jump to:
Product Details & Documents
Traditional rectangular coverslip style
- Quality slide making supplies
- Traditional coverslip type
Traditional style coverslips to use when making slides in the laboratory.
#1 Thickness: 0.15 mm thick. Approximately 75–100 coverslips per ounce.
#2 Thickness: 0.2 mm thick. Approximately 50-75 coverslips per ounce.
Especificaciones
- Numbering:#1
- Description:Glass cover slips, rectangular
- L×W:40×22 mm
- Thickness:0.15 mm