Order Entry
Mexico
Orders LinkContactUsLinkComponent
Square Glass Coverslips

Square Glass Coverslips

Glass cover slips, square, 18×18 mm, 0.15 mm, #1
Catalog # 470019-002
Número de proveedor #:  SCSS18-1OZ
Square Glass Coverslips
Catalog # 470019-002
Square Glass Coverslips
Square Glass Coverslips

Glass cover slips, square, 18×18 mm, 0.15 mm, #1

Price
$70.37
Log in Para disponibilidad
Paquete de 1
Product Details & Documents

Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.

  • Non corrosive glass
  • Pre-cleaned

Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.

Documents

Especificaciones
  • Numbering:
    #1
  • Description:
    Glass cover slips, square
  • L×W:
    18×18 mm
  • Thickness:
    0.15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.